Assay Details
| Gene Symbol: C12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152319.3 | NP_689532.1 | 2 - 3 | 198 | 81 | |
| GenBank mRNA | AY255793.1 | - | 1 - 2 | 103 | 81 | |
| BC031670.2 | - | 1 - 2 | 124 | 5165130 | 81 | |
| BQ429029.1 | - | 2 - 3 | 201 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000351.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000494130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004389.hg.1 | ENST00000314014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004389.hg.1 | ENST00000548364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004389.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004389.hg.1 | TC12000351.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004388.hg.1 | ENST00000314014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004388.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004388.hg.1 | TC12000351.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004390.hg.1 | ENST00000314014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004390.hg.1 | ENST00000548364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004390.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004390.hg.1 | TC12000351.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200152862.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152862.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152862.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152862.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152862.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152862.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152862.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152862.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152862.hg.1 | TC1200007522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000494130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152858.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152858.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152858.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152858.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152858.hg.1 | TC1200007522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200152860.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152860.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152860.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152860.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152860.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152860.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152860.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152860.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152860.hg.1 | TC1200007522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200152861.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152861.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152861.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152861.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152861.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152861.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152861.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152861.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152861.hg.1 | TC1200007522.hg.1 | Clariom D TC |

C12orf54
Genomic Map