Assay Details
| Gene Symbol: C12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152319.3 | NP_689532.1 | 3 - 4 | 220 | 69 | |
| GenBank mRNA | AY255793.1 | - | 2 - 3 | 125 | 69 | |
| AY255794.1 | - | 2 - 3 | 125 | 69 | ||
| BC031670.2 | - | 2 - 3 | 146 | 5165130 | 69 | |
| BQ429029.1 | - | 3 - 4 | 223 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12004393.hg.1 | ENST00000314014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004393.hg.1 | ENST00000548364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004393.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004393.hg.1 | TC12000351.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000494130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000314014.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000380491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000494130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548364.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000548913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000549398.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | ENST00000551266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007522.hg.1 | NM_152319.3 | Refseq |

C12orf54
Genomic Map