| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002504.hg.1 |
ENST00000256858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002504.hg.1 |
ENST00000398130.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002504.hg.1 |
NM_020854.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18002504.hg.1 |
TC18000220.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
ENST00000256858.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
ENST00000398130.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
ENST00000586920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
ENST00000587725.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
ENST00000588446.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
NM_020854.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
TC1800007504.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
ENST00000256858.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
ENST00000398130.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
ENST00000586920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
ENST00000587725.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
ENST00000588446.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800151435.hg.1 |
NM_020854.3 |
Refseq |