Assay Details
| Gene Symbol: YIPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173834.3 | NP_776195.2 | 2 - 3 | 472 | 108 | |
| XM_011530923.2 | XP_011529225.1 | 2 - 3 | 880 | 108 | ||
| XM_011530924.1 | XP_011529226.1 | 2 - 3 | 254 | 108 | ||
| XM_017029450.1 | XP_016884939.1 | 2 - 3 | 254 | 108 | ||
| GenBank mRNA | AK095555.1 | - | 2 - 3 | 254 | 109 | |
| AK292835.1 | - | 2 - 3 | 214 | 108 | ||
| BC012469.1 | - | 2 - 3 | 210 | 4452699 | 108 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000360.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004663.hg.1 | ENST00000374622.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004663.hg.1 | ENST00000462683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004663.hg.1 | ENST00000470730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004663.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004663.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X004663.hg.1 | TC0X000360.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007501.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000374622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000374643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000451537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000462683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000462972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000470730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | ENST00000496576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | NM_001195214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | NM_173834.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151369.hg.1 | TC0X00007501.hg.1 | Clariom D TC |

YIPF6
Genomic Map