| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000476112.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
ENST00000476112.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021944.hg.1 |
ENST00000361384.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021944.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021944.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10021944.hg.1 |
TC10001673.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000476112.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
ENST00000476112.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011885.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000181343.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181343.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181343.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000181343.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000181343.hg.1 |
ENST00000361384.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181343.hg.1 |
ENST00000369305.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000181343.hg.1 |
NM_001271816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000181343.hg.1 |
NM_014881.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000181343.hg.1 |
TC1000011885.hg.1 |
Clariom D TC |