| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007033.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007033.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007033.hg.1 |
TC08000520.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008088.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155547.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155547.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155547.hg.1 |
ENST00000297265.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155547.hg.1 |
NM_152284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155547.hg.1 |
TC0800008088.hg.1 |
Clariom D TC |