| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023853.hg.1 |
ENST00000369393.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023853.hg.1 |
ENST00000428876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023853.hg.1 |
NM_014611.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023853.hg.1 |
TC06001944.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023854.hg.1 |
ENST00000369393.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023854.hg.1 |
ENST00000428876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023854.hg.1 |
NM_014611.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023854.hg.1 |
TC06001944.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600186517.hg.1 |
ENST00000369393.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186517.hg.1 |
ENST00000629399.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186517.hg.1 |
NM_014611.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600186517.hg.1 |
ENST00000369393.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186517.hg.1 |
ENST00000629399.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186517.hg.1 |
NM_014611.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600186517.hg.1 |
TC0600012551.hg.1 |
Clariom D TC |