| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11011384.hg.1 |
ENST00000325212.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11011384.hg.1 |
ENST00000344196.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11011384.hg.1 |
ENST00000411936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11011384.hg.1 |
ENST00000531700.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11011384.hg.1 |
NM_033395.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11011384.hg.1 |
TC11000908.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100164803.hg.1 |
ENST00000325212.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100164803.hg.1 |
ENST00000530425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100164803.hg.1 |
ENST00000531700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100164803.hg.1 |
NM_033395.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100164803.hg.1 |
ENST00000325212.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100164803.hg.1 |
ENST00000530425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100164803.hg.1 |
ENST00000531700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100164803.hg.1 |
NM_033395.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100164803.hg.1 |
TC1100008786.hg.1 |
Clariom D TC |