| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030696.hg.1 |
ENST00000378061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030696.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030696.hg.1 |
TC07001997.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030698.hg.1 |
ENST00000378061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030698.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030698.hg.1 |
TC07001997.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030700.hg.1 |
ENST00000378061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030700.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030700.hg.1 |
TC07001997.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000484196.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000484196.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000484196.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000484196.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000495685.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000495685.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000495685.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
ENST00000495685.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000484196.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000484196.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000484196.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000484196.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000495685.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000495685.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000495685.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
ENST00000495685.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012971.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193548.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193548.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193548.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193548.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193548.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193548.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193548.hg.1 |
TC0700012971.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700193550.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193550.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193550.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193550.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193550.hg.1 |
TC0700012971.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700193545.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193545.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193545.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193545.hg.1 |
ENST00000378061.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193545.hg.1 |
ENST00000483014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193545.hg.1 |
NM_001001661.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193545.hg.1 |
TC0700012971.hg.1 |
Clariom D TC |