Assay Details
| Gene Symbol: C5orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198566.3 | NP_940968.1 | 10 - 11 | 1983 | 94 | |
| XM_011514038.2 | XP_011512340.1 | 10 - 11 | 1892 | 94 | ||
| XM_017009445.1 | XP_016864934.1 | 9 - 10 | 2014 | 94 | ||
| XM_017009446.1 | XP_016864935.1 | 8 - 9 | 1924 | 94 | ||
| GenBank mRNA | AK056925.1 | - | 10 - 11 | 1898 | 94 | |
| BC036867.1 | - | 10 - 11 | 1846 | 5198039 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001319.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000503655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000503655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000503655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000503655.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000509489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000509489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000509489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000509489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05018641.hg.1 | ENST00000306862.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05018641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05018641.hg.1 | TC05001319.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000504469.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007298.hg.1 | ENST00000505645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174322.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174322.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174322.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500174322.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174322.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500174322.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500174322.hg.1 | TC0500010641.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000306862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000503655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000503655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000503655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000503655.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000506213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000509489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000509489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000509489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000509489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | ENST00000514462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010641.hg.1 | NM_198566.3 | Refseq |

C5orf34
Genomic Map