Assay Details
| Gene Symbol: NUP210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024923.3 | NP_079199.2 | 34 - 35 | 4861 | 73 | |
| XM_017006018.1 | XP_016861507.1 | 16 - 17 | 2694 | 73 | ||
| XM_017006019.1 | XP_016861508.1 | 16 - 17 | 2661 | 73 | ||
| XM_017006020.1 | XP_016861509.1 | 16 - 17 | 4733 | 73 | ||
| GenBank mRNA | AB020713.1 | - | 14 - 15 | 1877 | 73 | |
| AK074101.1 | - | 19 - 20 | 2483 | 73 | ||
| AK075545.1 | - | 4 - 5 | 556 | 73 | ||
| AL117527.1 | - | 3 - 4 | 408 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001188.hg.1
Set Membership: |
|

NUP210
Genomic Map