Assay Details
| Gene Symbol: NHLRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198514.3 | NP_940916.2 | 1 - 2 | 415 | 64 | |
| XM_011539769.2 | XP_011538071.1 | 1 - 2 | 415 | 64 | ||
| GenBank mRNA | AK090631.1 | - | 1 - 2 | 386 | 64 | |
| DA821295.1 | - | 1 - 2 | 416 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000828.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009987.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009987.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009987.hg.1 | TC10000828.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009989.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009989.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009989.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009989.hg.1 | TC10000828.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009988.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009988.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009988.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009988.hg.1 | TC10000828.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000161304.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161304.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161304.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161304.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161304.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161304.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161304.hg.1 | TC1000008961.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000161302.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161302.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161302.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161302.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161302.hg.1 | TC1000008961.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000161305.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161305.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161305.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161305.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161305.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161305.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161305.hg.1 | TC1000008961.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008961.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161303.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161303.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161303.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161303.hg.1 | ENST00000369301.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161303.hg.1 | ENST00000468890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161303.hg.1 | NM_198514.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161303.hg.1 | TC1000008961.hg.1 | Clariom D TC |

NHLRC2
Genomic Map