Assay Details
| Gene Symbol: RTP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173821.2 | NP_776182.2 | 1 - 2 | 182 | 67 | |
| XM_011510997.2 | XP_011509299.1 | 1 - 2 | 187 | 67 | ||
| GenBank mRNA | AK090909.1 | - | 1 - 2 | 187 | 67 | |
| BC113517.1 | - | 1 - 2 | 182 | 8322569 | 67 | |
| BC113519.1 | - | 1 - 2 | 182 | 8322571 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001489.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138761 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138722 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138710 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138760 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138747 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138749 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138745 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138822 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138767 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138759 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138726 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138755 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138765 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138827 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138809 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138756 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138816 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138714 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138750 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138807 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138799 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138741 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138824 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138758 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138733 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138826 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138723 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22138729 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 550621 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P197284 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P197284 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 41524 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P102642 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P102642 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56698 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P13704977 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P197284 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P197284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138749 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138729 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138711 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138716 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138743 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138729 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138743 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138711 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138749 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138716 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22138733 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P197284 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P102642 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P102642 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03808408 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03808411 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P197284 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13704977 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_173821_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8049956 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8049953 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8049955 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8049954 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8049952 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1560035_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11735509_at | - | - |
| Affymetrix Human Genome U219 Array | 11735509_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024399.hg.1 | TC02001489.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024399.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024399.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024397.hg.1 | TC02001489.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024397.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02024397.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011431.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200182901.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182901.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182901.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200182901.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182901.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182901.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200182901.hg.1 | TC0200011431.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200182902.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182902.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182902.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200182902.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182902.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182902.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200182902.hg.1 | TC0200011431.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011431.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200182899.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182899.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182899.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200182899.hg.1 | ENST00000343216.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182899.hg.1 | ENST00000419912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200182899.hg.1 | NM_173821.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200182899.hg.1 | TC0200011431.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1560035_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1560035_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_173821_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560035_at5 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017224 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017224 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 205678 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1717029 | AK090909.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1717029 | AK090909.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1717029 | AK090909.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1717029 | AK090909.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1717029 | AK090909.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_28376647-S | AK090909.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_625-A | - | - |
| Illumina human-6 v1.0 expression beadchip | 0001340139 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0001340139 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1717029 | AK090909.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1717029 | AK090909.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1770386 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_625 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1717029 | AK090909.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1717029 | AK090909.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1717029 | AK090909.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1717029 | AK090909.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1717029 | AK090909.1 | human_transcriptome |

RTP5
Genomic Map