Assay Details
| Gene Symbol: SLC35E2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110781.2 | NP_001104251.1 | 7 - 8 | 1339 | 89 | |
| NM_001290264.1 | NP_001277193.1 | 8 - 9 | 1757 | 89 | ||
| GenBank mRNA | AB007916.3 | - | 7 - 8 | 1277 | 89 | |
| AK092311.1 | - | 1 - 2 | 478 | 89 | ||
| AK125619.1 | - | 7 - 8 | 1263 | 89 | ||
| BC110653.1 | - | 7 - 8 | 1315 | 4392812 | 89 | |
| CR936618.1 | - | 8 - 9 | 1477 | 89 | ||
| DA794179.1 | - | 4 - 5 | 393 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006344.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK092311.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032244.hg.1 | ENST00000234800.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032244.hg.1 | ENST00000378662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032244.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032244.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032244.hg.1 | TC01006344.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032247.hg.1 | ENST00000234800.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032247.hg.1 | ENST00000378662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032247.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032247.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032247.hg.1 | TC01006344.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032243.hg.1 | ENST00000234800.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032243.hg.1 | ENST00000378662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032243.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032243.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01032243.hg.1 | TC01006344.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241069.hg.1 | TC0100018388.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241070.hg.1 | TC0100018388.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100241072.hg.1 | TC0100018388.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000480991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000481276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000611123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000614300.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | ENST00000617444.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001110781.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018388.hg.1 | NM_001290264.1 | Refseq |

SLC35E2B
Genomic Map