| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010575.hg.1 |
ENST00000368656.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010575.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010575.hg.1 |
TC06000895.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010574.hg.1 |
ENST00000368656.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010574.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010574.hg.1 |
TC06000895.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010576.hg.1 |
ENST00000368656.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010576.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010576.hg.1 |
TC06000895.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010572.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010572.hg.1 |
TC06000895.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600162973.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162973.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162973.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162973.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162973.hg.1 |
TC0600009169.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009169.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162971.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162971.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162971.hg.1 |
TC0600009169.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600162978.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162978.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162978.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162978.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162978.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162978.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162978.hg.1 |
TC0600009169.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600162976.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162976.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162976.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162976.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162976.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162976.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162976.hg.1 |
TC0600009169.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600162974.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162974.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162974.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162974.hg.1 |
ENST00000368656.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162974.hg.1 |
ENST00000604268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162974.hg.1 |
NM_001033564.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162974.hg.1 |
TC0600009169.hg.1 |
Clariom D TC |