Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000528.hg.1 |
TC09000040.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000528.hg.1 |
ENST00000251879.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000528.hg.1 |
ENST00000381532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000528.hg.1 |
ENST00000414202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000528.hg.1 |
ENST00000418622.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000528.hg.1 |
ENST00000449720.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000528.hg.1 |
NM_001135920.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000528.hg.1 |
NM_001206557.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09000528.hg.1 |
NM_020829.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
ENST00000251879.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
ENST00000276898.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
ENST00000414202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
ENST00000418622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
ENST00000545641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
NM_001135920.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
NM_001206557.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
NM_020829.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
ENST00000251879.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
ENST00000276898.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
ENST00000414202.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
ENST00000418622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
ENST00000545641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
NM_001135920.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
NM_001206557.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
NM_020829.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900144942.hg.1 |
TC0900006564.hg.1 |
Clariom D TC |