Assay Details
| Gene Symbol: SIAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199922.1 | NP_001186851.1 | 8 - 9 | 1131 | 86 | |
| NM_170601.4 | NP_733746.1 | 6 - 7 | 1000 | 86 | ||
| XM_017017930.1 | XP_016873419.1 | 7 - 8 | 3520 | 86 | ||
| GenBank mRNA | AF300796.1 | - | 8 - 9 | 1131 | 86 | |
| AF303378.1 | - | 6 - 7 | 839 | 86 | ||
| AK056093.1 | - | 6 - 7 | 866 | 86 | ||
| AK092224.1 | - | 2 - 3 | 179 | 86 | ||
| AL137496.1 | - | 6 - 7 | 822 | 86 | ||
| BC022456.1 | - | 6 - 7 | 854 | 86 | ||
| BC068450.1 | - | 6 - 7 | 986 | 5278370 | 86 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002410.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11028843.hg.1 | ENST00000263593.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028843.hg.1 | ENST00000545756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028843.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028843.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028843.hg.1 | TC11002410.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000436137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000525730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000533613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012681.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | ENST00000263593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | ENST00000545756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | ENST00000618733.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | NM_001199922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | NM_170601.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100197452.hg.1 | TC1100012681.hg.1 | Clariom D TC |

SIAE
Genomic Map