Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011127.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011127.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011127.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011127.hg.1 |
TC10000909.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000508096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000508096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
ENST00000508096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163238.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163238.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163238.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163238.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163238.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000163238.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000163238.hg.1 |
TC1000009221.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000359653.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000471421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000508096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000508096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
ENST00000508096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009221.hg.1 |
NM_017580.2 |
Refseq |