| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000455386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000455386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000455386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000455386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000465048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000465048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
ENST00000465048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009259.hg.1 |
ENST00000369777.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009259.hg.1 |
ENST00000369780.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009259.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10009259.hg.1 |
TC10000775.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000160108.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160108.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160108.hg.1 |
ENST00000455386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160108.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000160108.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160108.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160108.hg.1 |
ENST00000455386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000160108.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000160108.hg.1 |
TC1000008776.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000369780.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000437579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000455386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000455386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000455386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000455386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000465048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000465048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
ENST00000465048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008776.hg.1 |
NM_004210.4 |
Refseq |