Assay Details
| Gene Symbol: FMNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052905.3 | NP_443137.2 | 8 - 9 | 1154 | 63 | |
| XM_005246263.2 | XP_005246320.1 | 8 - 9 | 1221 | 63 | ||
| XM_005246265.3 | XP_005246322.1 | 8 - 9 | 1221 | 63 | ||
| XM_011510530.1 | XP_011508832.1 | 8 - 9 | 1221 | 63 | ||
| XM_011510531.1 | XP_011508833.1 | 8 - 9 | 1221 | 63 | ||
| XM_011510532.2 | XP_011508834.1 | 8 - 9 | 1221 | 63 | ||
| XM_011510533.2 | XP_011508835.1 | 8 - 9 | 1221 | 63 | ||
| XM_011510534.1 | XP_011508836.1 | 8 - 9 | 1221 | 63 | ||
| XM_011510535.2 | XP_011508837.1 | 7 - 8 | 1068 | 63 | ||
| XM_011510536.2 | XP_011508838.1 | 2 - 3 | 172 | 63 | ||
| GenBank mRNA | AB067489.1 | - | 8 - 9 | 922 | 63 | |
| AL834396.1 | - | 6 - 7 | 535 | 63 | ||
| BC167159.1 | - | 8 - 9 | 922 | 8862581 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000941.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02013754.hg.1 | ENST00000288670.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013754.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013754.hg.1 | TC02000941.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000497192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000497192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | ENST00000497192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013755.hg.1 | ENST00000288670.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013755.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013755.hg.1 | TC02000941.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200167649.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167649.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167649.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167649.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167649.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167649.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167649.hg.1 | TC0200009680.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200167647.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167647.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167647.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167647.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167647.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167647.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167647.hg.1 | TC0200009680.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000288670.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000475377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000492942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000497192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000497192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | ENST00000497192.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009680.hg.1 | NM_052905.3 | Refseq |

FMNL2
Genomic Map