| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008234.hg.1 |
ENST00000336273.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008234.hg.1 |
ENST00000519934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008234.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008234.hg.1 |
TC08000594.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
ENST00000522313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008314.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
ENST00000522104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
ENST00000336273.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
ENST00000519293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
ENST00000519934.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
ENST00000521933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
ENST00000522104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
NM_178812.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157258.hg.1 |
TC0800008314.hg.1 |
Clariom D TC |