| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006883.hg.1 |
ENST00000257632.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006883.hg.1 |
ENST00000334348.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006883.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006883.hg.1 |
ENST00000419923.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006883.hg.1 |
ENST00000443097.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006883.hg.1 |
ENST00000448265.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006883.hg.1 |
NM_030570.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006883.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006883.hg.1 |
TC07003310.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
ENST00000469114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
ENST00000469114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156204.hg.1 |
TC0700008096.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000257632.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000334348.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000394849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000447046.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000469114.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
ENST00000490360.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008096.hg.1 |
NM_182684.2 |
Refseq |