Assay Details
| Gene Symbol: LINC01213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110167.1 | 1 - 2 | 131 | 97 | ||
| GenBank mRNA | BF676681.1 | - | 1 - 2 | 132 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009157.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165650.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165650.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165650.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165650.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165650.hg.1 | TC0300009157.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | ENST00000472890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | NR_110168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165653.hg.1 | TC0300009157.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300165651.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165651.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165651.hg.1 | ENST00000498005.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300165651.hg.1 | NR_110167.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300165651.hg.1 | TC0300009157.hg.1 | Clariom D TC |

LINC01213
Genomic Map