Assay Details
| Gene Symbol: LINC01208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109968.1 | 2 - 3 | 283 | 129 | ||
| GenBank mRNA | BX094192.1 | - | 2 - 3 | 283 | 129 | |
| CA436377.1 | - | 1 - 2 | 129 | |||
More Information
Set Membership: |
|

LINC01208
Genomic Map