Assay Details
| Gene Symbol: TMEM247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145051.2 | NP_001138523.1 | 2 - 3 | 496 | 93 | |
| GenBank mRNA | AA432195.1 | - | 1 - 2 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02004962.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 172327 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 838152 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 18691 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 363789 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 55981 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 325609 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 145521 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 146719 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 212143 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3664161 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1977 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1976 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60215 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3664161 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3664161 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152523 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152532 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21233021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152533 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152536 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21233001 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152519 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152538 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152543 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21233011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21233022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21233011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21233001 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152533 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152519 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152532 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152538 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21233021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21233022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152543 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152536 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152523 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01152525 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15240157 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152610 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152512 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152556 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152573 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152585 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15240151 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15240156 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152626 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152541 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152608 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152591 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152538 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152525 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152611 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152598 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152566 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152519 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152516 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152576 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152614 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152594 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152575 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152545 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152532 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152602 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152543 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152620 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152507 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152587 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152549 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152536 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152618 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152547 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152568 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152600 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152622 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152510 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15240161 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152570 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152583 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152521 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152580 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15240152 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15240155 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152522 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152606 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152604 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152527 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152589 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152562 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152613 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152552 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01152514 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P13197807 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8041806 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8041807 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8041805 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8041804 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1556490_PM_a_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1556489_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11738359_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11738359_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004302.hg.1 | ENST00000434431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004302.hg.1 | NM_001145051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004302.hg.1 | TC02004962.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016441.hg.1 | ENST00000434431.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016441.hg.1 | ENST00000434431.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016441.hg.1 | ENST00000434431.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016441.hg.1 | NM_001145051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016441.hg.1 | NM_001145051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016441.hg.1 | NM_001145051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221673.hg.1 | ENST00000434431.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221673.hg.1 | NM_001145051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221673.hg.1 | ENST00000434431.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221673.hg.1 | NM_001145051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221673.hg.1 | TC0200016441.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016441.hg.1 | ENST00000434431.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016441.hg.1 | ENST00000434431.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016441.hg.1 | ENST00000434431.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016441.hg.1 | NM_001145051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016441.hg.1 | NM_001145051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016441.hg.1 | NM_001145051.2 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1556490_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1556489_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1556490_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1556489_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556489_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1556490_a_at1 | - | - |
| Human Genome U95 Set | 42190_PM_at | - | - |
| Human Genome U95 Set | 42190_at | - | - |
| Human Genome U95 Set | 50465_at | - | - |
| Human Genome U95 Set | 50465_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015606 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015606 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 178632 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1752257 | NM_001145051.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1752257 | AA432195.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1752257 | NM_001145051.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1752257 | AA432195.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1752257 | NM_001145051.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1752257 | AA432195.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1752257 | NM_001145051.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1752257 | AA432195.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1752257 | NM_001145051.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1752257 | AA432195.1 | human_transcriptome |

TMEM247
Genomic Map