Assay Details
| Gene Symbol: LOC101927000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132384.1 | 1 - 2 | 54 | 83 | ||
| GenBank mRNA | BX118161.1 | - | 1 - 2 | 54 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101927000
Genomic Map