| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009565.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009565.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009565.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009565.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009565.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009565.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009565.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200166782.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200166782.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200166782.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200166782.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200166782.hg.1 |
TC0200009565.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200166783.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200166783.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200166783.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200166783.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200166783.hg.1 |
TC0200009565.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200009565.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009565.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009565.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009565.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009565.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009565.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009565.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200166780.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200166780.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200166780.hg.1 |
TC0200009565.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200166784.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200166784.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200166784.hg.1 |
ENST00000413525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200166784.hg.1 |
NR_110570.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200166784.hg.1 |
TC0200009565.hg.1 |
Clariom D TC |