Assay Details
| Gene Symbol: LOC101927359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125885.1 | 7 - 8 | 584 | 139 | ||
| GenBank mRNA | BC040219.1 | - | 7 - 8 | 581 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181529.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181529.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181529.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181529.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181529.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181529.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181529.hg.1 | TC0400011870.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400181528.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181528.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181528.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181528.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181528.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181528.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181528.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181528.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181528.hg.1 | TC0400011870.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181525.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181525.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181525.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181525.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181525.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181525.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181525.hg.1 | TC0400011870.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400181527.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181527.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181527.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181527.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181527.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181527.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181527.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181527.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181527.hg.1 | TC0400011870.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400181526.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181526.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181526.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181526.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181526.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181526.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181526.hg.1 | TC0400011870.hg.1 | Clariom D TC |

LOC101927359
Genomic Map