Assay Details
| Gene Symbol: ILDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199799.1 | NP_001186728.1 | 3 - 4 | 587 | 67 | |
| NM_175924.3 | NP_787120.1 | 3 - 4 | 587 | 67 | ||
| XM_005247389.4 | XP_005247446.1 | 2 - 3 | 999 | 67 | ||
| XM_011512738.2 | XP_011511040.1 | 3 - 4 | 567 | 67 | ||
| XM_011512739.2 | XP_011511041.1 | 3 - 4 | 461 | 67 | ||
| GenBank mRNA | AY134857.1 | - | 3 - 4 | 505 | 67 | |
| AY672837.1 | - | 3 - 4 | 429 | 67 | ||
| AY672838.1 | - | 2 - 3 | 466 | 67 | ||
| BC044240.1 | - | 3 - 4 | 490 | 5751749 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001700.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011512739.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030772.hg.1 | ENST00000273691.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030772.hg.1 | ENST00000344209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030772.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030772.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030772.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030772.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030772.hg.1 | TC03001700.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030773.hg.1 | ENST00000273691.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030773.hg.1 | ENST00000344209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030773.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030773.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030773.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030773.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030773.hg.1 | TC03001700.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190712.hg.1 | TC0300012178.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190713.hg.1 | TC0300012178.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 162313 | - | - |

ILDR1
Genomic Map