Assay Details
| Gene Symbol: FAM159A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042693.2 | NP_001036158.1 | 1 - 2 | 329 | 62 | |
| NR_135157.1 | 1 - 2 | 329 | 62 | |||
| GenBank mRNA | AK128236.1 | - | 1 - 2 | 329 | 62 | |
| AY358621.1 | - | 1 - 2 | 103 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000644.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY358621.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010505.hg.1 | ENST00000401050.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010505.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010505.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010505.hg.1 | TC01000644.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010508.hg.1 | ENST00000401050.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010508.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010508.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010508.hg.1 | TC01000644.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010506.hg.1 | ENST00000401050.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010506.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010506.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010506.hg.1 | TC01000644.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100159297.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159297.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159297.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159297.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159297.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159297.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159297.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159297.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159297.hg.1 | TC0100008326.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008326.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159298.hg.1 | TC0100008326.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | ENST00000424164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159300.hg.1 | TC0100008326.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100159296.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159296.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159296.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159296.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159296.hg.1 | ENST00000401050.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159296.hg.1 | ENST00000440303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159296.hg.1 | ENST00000517870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159296.hg.1 | NM_001042693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159296.hg.1 | TC0100008326.hg.1 | Clariom D TC |

FAM159A
Genomic Map