Assay Details
| Gene Symbol: CDPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207327.4 | NP_997210.3 | 2 - 3 | 153 | 79 | |
| XM_011529960.1 | XP_011528262.1 | 2 - 3 | 536 | 79 | ||
| XM_011529962.2 | XP_011528264.1 | 2 - 3 | 144 | 79 | ||
| XM_011529965.2 | XP_011528267.1 | 2 - 3 | 469 | 79 | ||
| GenBank mRNA | BC036462.1 | - | 2 - 3 | 151 | 79 | |
| BC045796.2 | - | 1 - 2 | 83 | 79 | ||
| BC067871.1 | - | 2 - 3 | 153 | 30338205 | 79 | |
| BX648331.1 | - | 2 - 3 | 161 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000886.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014909.hg.1 | ENST00000314567.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014909.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014909.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014909.hg.1 | TC22000886.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014906.hg.1 | ENST00000314567.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014906.hg.1 | ENST00000404583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014906.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014906.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014906.hg.1 | TC22000886.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014907.hg.1 | ENST00000314567.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014907.hg.1 | ENST00000404583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014907.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014907.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014907.hg.1 | TC22000886.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | TC2200009013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165311.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | TC2200009013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165312.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | TC2200009013.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165314.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000314567.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000381037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000404744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000474256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | ENST00000475605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009013.hg.1 | NM_207327.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 136656 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 156348 | - | - |

CDPF1
Genomic Map