Assay Details
| Gene Symbol: PYY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003064.2 | 1 - 2 | 317 | 97 | ||
| GenBank mRNA | AF222904.1 | - | 1 - 2 | 66 | 97 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000317.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF222904.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10271260 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16965183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30835243 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10271271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30835247 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10271263 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10271264 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10271270 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P233078 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3257866 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P233078 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 38463 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 39738 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4434 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3257866 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P233078 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P233078 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3257866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965193 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271254 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271246 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271254 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271246 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965193 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271267 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P233078 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P233078 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271272 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271265 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271264 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271269 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271277 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271267 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271250 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16965195 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271254 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271275 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271274 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16965167 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271260 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271248 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16965186 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271252 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271271 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271258 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271256 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271270 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16965199 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003064_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8005834 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8005835 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8005833 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 220209_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11744736_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11744736_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000315776.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000315776.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003825.hg.1 | ENST00000441253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003825.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003825.hg.1 | TC17000317.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003823.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003823.hg.1 | TC17000317.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003824.hg.1 | ENST00000441253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003824.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17003824.hg.1 | TC17000317.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700152248.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152248.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152248.hg.1 | TC1700007355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152248.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152248.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152251.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152251.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152251.hg.1 | TC1700007355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152251.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152251.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152256.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152256.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152256.hg.1 | TC1700007355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152256.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152256.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000315776.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000315776.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152252.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152252.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152252.hg.1 | TC1700007355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152252.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152252.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152254.hg.1 | ENST00000315776.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152254.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152254.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152254.hg.1 | TC1700007355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152254.hg.1 | ENST00000315776.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152254.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152254.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152249.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152249.hg.1 | NR_003064.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152249.hg.1 | TC1700007355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152249.hg.1 | ENST00000441253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152249.hg.1 | NR_003064.2 | Refseq |
| Human Genome U133 Plus 2.0 Array | 220209_at | - | - |
| Human Genome U133 Plus 2.0 Array | 220209_PM_at | - | - |
| Human Genome U133A 2.0 Array | 220209_PM_at | - | - |
| Human Genome U133A 2.0 Array | 220209_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw523903 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at3 | - | - |
| Human Genome U95 Set | 83732_PM_at | - | - |
| Human Genome U95 Set | 83732_at | - | - |
| Human Genome U133 Set | 220209_at | - | - |
| Human Genome U133 Set | 220209_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009296 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009295 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009296 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009295 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 212372 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1699565 | AF222904.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2070395 | NR_003064.2 | human_transcriptome |

PYY2
Genomic Map