Assay Details
Gene Symbol: ST6GALNAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175039.3 | NP_778204.1 | 1 - 2 | 205 | 103 | |
XM_011518527.2 | XP_011516829.1 | 1 - 2 | 228 | 103 | ||
XM_011518529.2 | XP_011516831.1 | 1 - 2 | 231 | 103 | ||
GenBank mRNA | AJ271734.1 | - | 1 - 2 | 219 | 104 | |
AK000600.1 | - | 1 - 2 | 201 | 103 | ||
BC036705.2 | - | 1 - 2 | 111 | 4309208 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001615.hg.1
Additional Information:
This assay is in the 5'UTR region of the AJ271734.1,AK000600.1,BC036705.2,NM_175039.3,XM_011518527.2,XM_011518529.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
glycolipid metabolic process
oligosaccharide metabolic process
O-glycan processing
protein N-linked glycosylation via asparagine
sialylation
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
sialyltransferase activity
(alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl-galactosaminide 6-alpha-sialyltransferase activity
ganglioside biosynthetic process
glycolipid metabolic process
oligosaccharide metabolic process
O-glycan processing
protein N-linked glycosylation via asparagine
sialylation
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
sialyltransferase activity
(alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl-galactosaminide 6-alpha-sialyltransferase activity
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09020365.hg.1 | TC09001615.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09020365.hg.1 | ENST00000335791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020365.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09020367.hg.1 | TC09001615.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09020367.hg.1 | ENST00000335791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020367.hg.1 | ENST00000343609.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020367.hg.1 | ENST00000541933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020367.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09020367.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09020366.hg.1 | TC09001615.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09020366.hg.1 | ENST00000335791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020366.hg.1 | ENST00000343609.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020366.hg.1 | ENST00000541933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09020366.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09020366.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179220.hg.1 | TC0900011610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179219.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179219.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179219.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179219.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179219.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179219.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179219.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179219.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179219.hg.1 | TC0900011610.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900179221.hg.1 | TC0900011610.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 123115 | - | - |
