Assay Details
| Gene Symbol: NHLRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK000154.1 | - | 1 | 254 | 67 | |
| AK126751.1 | - | 1 | 1301 | 67 | ||
| BC013743.1 | - | 1 | 718 | 67 | ||
| BX647663.1 | - | 1 | 1806 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NHLRC2
Genomic Map