Assay Details
| Gene Symbol: LBX2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024606.1 | 1 | 1489 | 78 | ||
| GenBank mRNA | AK096725.1 | - | 1 | 920 | 78 | |
| AK125271.1 | - | 1 | 1489 | 78 | ||
| BC009264.2 | - | 1 | 942 | 78 | ||
| BC101224.2 | - | 1 | 381 | 78 | ||
| BC101225.2 | - | 1 | 381 | 78 | ||
| BC101226.1 | - | 1 | 381 | 78 | ||
| BC101227.1 | - | 1 | 381 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000461.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 101333 | AK025844.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 101333 | BC004255.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 101333 | NM_025231.1 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 51990 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P319640 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P319640 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 34045 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P107767 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P107767 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42444 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P319640 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P319640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281663 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21363817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281683 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281661 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281654 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21363823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21363824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281665 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21363791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281685 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21363817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281651 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281665 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281663 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281683 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21363791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281685 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21363824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281661 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21363823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281654 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01281669 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P319640 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P107767 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P107767 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P107767 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P319640 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 72013 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 95384 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P107767 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281727 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281646 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15257288 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281752 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281643 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281672 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281655 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281656 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281654 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281683 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281663 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281753 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281658 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281677 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15257284 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281661 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281648 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281685 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281720 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281729 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281674 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15257295 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281666 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281758 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281679 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281756 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281718 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15257311 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281716 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281754 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281669 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281644 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01281652 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P107767 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 238422_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | ENST00000548978.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | ENST00000548978.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | ENST00000548978.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | ENST00000548978.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007204.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007204.hg.1 | TC02000461.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200156115.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156115.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156115.hg.1 | TC0200008087.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200156112.hg.1 | ENST00000548978.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156112.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156112.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156112.hg.1 | ENST00000548978.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156112.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156112.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156112.hg.1 | TC0200008087.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | ENST00000548978.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | ENST00000548978.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | ENST00000548978.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | ENST00000548978.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | ENST00000603175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008087.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156114.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156114.hg.1 | NR_024606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156114.hg.1 | TC0200008087.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 238422_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 238422_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 238422_at3 | - | - |
| Human Genome U95 Set | 68693_PM_at | - | - |
| Human Genome U95 Set | 68693_at | - | - |
| Human Genome U133 Set | 238422_PM_at | - | - |
| Human Genome U133 Set | 238422_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015855 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015856 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015856 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015855 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 214313 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3249231 | AK125271.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3249231 | NR_024606.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3249231 | AK096725.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3249231 | AK125271.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3249231 | NR_024606.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3249231 | AK096725.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3249231 | AK125271.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3249231 | NR_024606.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3249231 | AK096725.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3249231 | AK125271.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3249231 | NR_024606.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3249231 | AK096725.1 | human_transcriptome |

LBX2-AS1
Genomic Map