| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000229563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000466421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000466421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000466421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000466421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000466421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000466421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000466421.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000467415.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000467415.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000467415.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000467415.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000467415.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000467415.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000467415.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000467415.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000467415.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000495549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000495549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000495549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000495549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000495549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
ENST00000541412.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_001165258.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014068.hg.1 |
NM_016462.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010794.hg.1 |
ENST00000606522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010794.hg.1 |
ENST00000606522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010794.hg.1 |
ENST00000606652.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010794.hg.1 |
ENST00000606652.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010794.hg.1 |
ENST00000606652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010794.hg.1 |
ENST00000606522.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010794.hg.1 |
ENST00000606522.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010794.hg.1 |
ENST00000606652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010794.hg.1 |
ENST00000606652.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010794.hg.1 |
ENST00000606652.1 |
Ensembl |