Assay Details
| Gene Symbol: PGAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138575.3 | NP_612642.2 | 5 - 6 | 789 | 64 | |
| GenBank mRNA | BC008196.1 | - | 5 - 6 | 746 | 3048106 | 64 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003265.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12014054.hg.1 | ENST00000317555.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014054.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014054.hg.1 | TC12003265.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202137.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202137.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202137.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202137.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202137.hg.1 | TC1200012731.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000454808.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000498926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | ENST00000543955.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_001170544.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012731.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202138.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202138.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202138.hg.1 | ENST00000317555.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202138.hg.1 | NM_138575.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202138.hg.1 | TC1200012731.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 138197 | - | - |

PGAM5
Genomic Map