| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018080.hg.1 |
ENST00000396393.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018080.hg.1 |
ENST00000396394.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018080.hg.1 |
ENST00000418988.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018080.hg.1 |
ENST00000530457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018080.hg.1 |
NM_001143937.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018080.hg.1 |
NM_002786.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018080.hg.1 |
NM_148976.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018080.hg.1 |
TC11003474.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000396394.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000418988.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000526443.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000528307.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000530457.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000531156.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000533068.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000533331.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
NM_001143937.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
NM_002786.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
NM_148976.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000396394.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000418988.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000526443.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000528307.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000530457.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000531156.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000533068.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
ENST00000533331.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
NM_001143937.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
NM_002786.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
NM_148976.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203633.hg.1 |
TC1100013146.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000396394.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000418988.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000526443.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000528307.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000530457.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000531156.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000532256.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000533068.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000533331.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
NM_001143937.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
NM_002786.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
NM_148976.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000396394.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000418988.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000526443.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000528307.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000530457.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000531156.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000532256.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000533068.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
ENST00000533331.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
NM_001143937.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
NM_002786.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
NM_148976.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203634.hg.1 |
TC1100013146.hg.1 |
Clariom D TC |