| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012441.hg.1 |
ENST00000324382.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012441.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012441.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03012441.hg.1 |
TC03000694.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162530.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162530.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162530.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162530.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162530.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162530.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300162530.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162530.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300162530.hg.1 |
TC0300008789.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
ENST00000324382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
ENST00000503977.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_001308262.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008789.hg.1 |
NM_153833.2 |
Refseq |