Assay Details
| Gene Symbol: GPBAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170699.2 | NP_733800.1 | 1 - 2 | 198 | 91 | |
| XM_017003467.1 | XP_016858956.1 | 2 - 3 | 1423 | 91 | ||
| GenBank mRNA | BC033625.1 | - | 1 - 2 | 182 | 5221127 | 91 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001296.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC033625.1,NM_170699.2,XM_017003467.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02019775.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019775.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019775.hg.1 | ENST00000522678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019775.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019775.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019775.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019775.hg.1 | TC02001296.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010790.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019781.hg.1 | ENST00000479077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019781.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019781.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019781.hg.1 | ENST00000522678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019781.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019781.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019781.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019781.hg.1 | TC02001296.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019780.hg.1 | ENST00000479077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019780.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019780.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019780.hg.1 | ENST00000522678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019780.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019780.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019780.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019780.hg.1 | TC02001296.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019777.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019777.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019777.hg.1 | ENST00000522678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019777.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019777.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019777.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02019777.hg.1 | TC02001296.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176615.hg.1 | TC0200010790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176616.hg.1 | TC0200010790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200176612.hg.1 | TC0200010790.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000479077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000519574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000521462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | ENST00000522678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077191.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_001077194.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_170699.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010790.hg.1 | NM_170699.2 | Refseq |

GPBAR1
Genomic Map