Assay Details
| Gene Symbol: IL1RL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104167.1 | 6 - 7 | 1022 | 109 | ||
| GenBank mRNA | AB029084.1 | - | 6 - 7 | 984 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL1RL1| Gene Symbol: IL1RL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104167.1 | 6 - 7 | 1022 | 109 | ||
| GenBank mRNA | AB029084.1 | - | 6 - 7 | 984 | 109 | |
Set Membership: |
|