Assay Details
| Gene Symbol: BPIFB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174897.2 | NP_777557.1 | 6 - 7 | 584 | 66 | |
| XM_017027663.1 | XP_016883152.1 | 6 - 7 | 632 | 66 | ||
| GenBank mRNA | AF465767.1 | - | 6 - 7 | 584 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000221.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20002751.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002751.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20002751.hg.1 | TC20000221.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148874.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148874.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148874.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000148874.hg.1 | TC2000007139.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000148874.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148874.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000148874.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000349552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | ENST00000542375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007139.hg.1 | NM_174897.2 | Refseq |

BPIFB6
Genomic Map