Assay Details
Gene Symbol: ST6GALNAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK292049.1 | - | 5 - 6 | 1156 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
