| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014525.hg.1 |
ENST00000344229.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014525.hg.1 |
ENST00000361785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014525.hg.1 |
ENST00000392894.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014525.hg.1 |
ENST00000543506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014525.hg.1 |
NM_007282.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014525.hg.1 |
NM_183381.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014525.hg.1 |
TC03000809.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000344229.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000361785.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000392894.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000459632.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000466795.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000467977.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000467996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000474348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000482083.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000491086.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
NM_007282.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
NM_183381.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000344229.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000361785.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000392894.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000459632.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000466795.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000467977.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000467996.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000474348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000482083.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
ENST00000491086.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
NM_007282.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
NM_183381.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300165596.hg.1 |
TC0300009147.hg.1 |
Clariom D TC |