Assay Details
| Gene Symbol: TIPARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184717.1 | NP_001171646.1 | 3 - 4 | 1593 | 67 | |
| NM_001184718.1 | NP_001171647.1 | 3 - 4 | 1158 | 67 | ||
| NM_015508.4 | NP_056323.2 | 3 - 4 | 1508 | 67 | ||
| GenBank mRNA | AK097785.1 | - | 3 - 4 | 1337 | 67 | |
| AK291261.1 | - | 3 - 4 | 1343 | 67 | ||
| AL080156.1 | - | 2 - 3 | 194 | 67 | ||
| BC034397.1 | - | 3 - 4 | 1158 | 67 | ||
| BC050350.1 | - | 3 - 4 | 1315 | 5264807 | 67 | |
| BX537965.1 | - | 3 - 4 | 1335 | 67 | ||
| CR749647.1 | - | 3 - 4 | 1594 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000853.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
vasculogenesis
kidney development
protein ADP-ribosylation
nitrogen compound metabolic process
androgen metabolic process
estrogen metabolic process
female gonad development
post-embryonic development
negative regulation of gene expression
hemopoiesis
multicellular organism metabolic process
positive regulation of protein catabolic process
platelet-derived growth factor receptor signaling pathway
skeletal system morphogenesis
smooth muscle tissue development
palate development
face morphogenesis
cellular response to organic cyclic compound
NAD+ ADP-ribosyltransferase activity
enhancer binding
metal ion binding
vasculogenesis
kidney development
protein ADP-ribosylation
nitrogen compound metabolic process
androgen metabolic process
estrogen metabolic process
female gonad development
post-embryonic development
negative regulation of gene expression
hemopoiesis
multicellular organism metabolic process
positive regulation of protein catabolic process
platelet-derived growth factor receptor signaling pathway
skeletal system morphogenesis
smooth muscle tissue development
palate development
face morphogenesis
cellular response to organic cyclic compound
NAD+ ADP-ribosyltransferase activity
enhancer binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03015106.hg.1 | ENST00000295924.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015106.hg.1 | ENST00000461166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015106.hg.1 | ENST00000486483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015106.hg.1 | ENST00000542783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015106.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015106.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015106.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015106.hg.1 | TC03000853.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300166529.hg.1 | TC0300009282.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000295924.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000461166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000473702.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000481853.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000486483.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000495891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | ENST00000542783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_001184718.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_015508.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009282.hg.1 | NM_015508.4 | Refseq |

TIPARP
Genomic Map