| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022981.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022981.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022981.hg.1 |
TC02001437.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022983.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022983.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022983.hg.1 |
TC02001437.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022984.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022984.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022984.hg.1 |
TC02001437.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022980.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022980.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02022980.hg.1 |
TC02001437.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200180875.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180875.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200180875.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180875.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200180875.hg.1 |
TC0200011219.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200180879.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180879.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180879.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200180879.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180879.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180879.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200180879.hg.1 |
TC0200011219.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011219.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200180874.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180874.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200180874.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180874.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200180874.hg.1 |
TC0200011219.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200180877.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180877.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180877.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200180877.hg.1 |
ENST00000272928.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180877.hg.1 |
ENST00000447924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200180877.hg.1 |
NM_020311.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200180877.hg.1 |
TC0200011219.hg.1 |
Clariom D TC |