| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049384.hg.1 |
ENST00000271688.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049384.hg.1 |
ENST00000345896.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049384.hg.1 |
ENST00000368954.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049384.hg.1 |
ENST00000560793.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049384.hg.1 |
ENST00000561294.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049384.hg.1 |
NM_022075.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049384.hg.1 |
NM_181746.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049384.hg.1 |
TC01003215.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000271688.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000345896.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000361419.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000368949.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000368954.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000421609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000460664.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000558062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000559020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000559660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000560793.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000561294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
NM_022075.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
NM_181746.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000271688.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000345896.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000361419.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000368949.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000368954.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000421609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000460664.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000558062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000559020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000559660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000560793.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
ENST00000561294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
NM_022075.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
NM_181746.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218136.hg.1 |
TC0100015761.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000271688.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000345896.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000361419.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000368949.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000368954.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000421609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000460664.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000482825.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000558062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000559020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000559660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000560793.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000561294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
NM_022075.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
NM_181746.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000271688.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000345896.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000361419.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000368949.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000368954.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000421609.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000460664.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000482825.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000558062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000559020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000559660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000560793.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
ENST00000561294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
NM_022075.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
NM_181746.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218133.hg.1 |
TC0100015761.hg.1 |
Clariom D TC |