| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016153.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016153.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016153.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016153.hg.1 |
TC17001187.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016152.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016152.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016152.hg.1 |
TC17001187.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016151.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016151.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016151.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016151.hg.1 |
TC17001187.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700177698.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177698.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177698.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177698.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177698.hg.1 |
TC1700009938.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700177698.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177698.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177698.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177698.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009938.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177699.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177699.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177699.hg.1 |
TC1700009938.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700177699.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177699.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177700.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177700.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177700.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177700.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177700.hg.1 |
TC1700009938.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700177700.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177700.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177700.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177700.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177697.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177697.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177697.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177697.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177697.hg.1 |
TC1700009938.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700177697.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177697.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177697.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177697.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177701.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177701.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177701.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177701.hg.1 |
NM_016084.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177701.hg.1 |
TC1700009938.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700177701.hg.1 |
ENST00000225688.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177701.hg.1 |
ENST00000579152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177701.hg.1 |
NM_001199989.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177701.hg.1 |
NM_016084.4 |
Refseq |