Assay Details
| Gene Symbol: WNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003391.2 | NP_003382.1 | 2 - 3 | 614 | 82 | |
| GenBank mRNA | AK056742.1 | - | 2 - 3 | 611 | 82 | |
| BC029854.1 | - | 2 - 3 | 366 | 82 | ||
| BC078170.1 | - | 2 - 3 | 600 | 5761421 | 82 | |
| BT019608.1 | - | 2 - 3 | 314 | 82 | ||
| X07876.1 | - | 2 - 3 | 608 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001794.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of endothelial cell proliferation
positive regulation of mesenchymal cell proliferation
lens development in camera-type eye
cell-cell signaling
positive regulation of cell proliferation
Wnt signaling pathway
neurogenesis
neuron differentiation
lung development
cell proliferation in midbrain
cell fate commitment
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fibroblast proliferation
positive regulation of neurogenesis
positive regulation of sequence-specific DNA binding transcription factor activity
atrial cardiac muscle tissue morphogenesis
positive regulation of cardiac muscle cell proliferation
canonical Wnt signaling pathway
cardiac epithelial to mesenchymal transition
lung induction
positive regulation of epithelial cell proliferation involved in lung morphogenesis
labyrinthine layer blood vessel development
iris morphogenesis
mammary gland epithelium development
cellular response to retinoic acid
cellular response to transforming growth factor beta stimulus
positive regulation of canonical Wnt signaling pathway
midbrain dopaminergic neuron differentiation
canonical Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
frizzled binding
cytokine activity
protein binding
receptor agonist activity
positive regulation of endothelial cell proliferation
positive regulation of mesenchymal cell proliferation
lens development in camera-type eye
cell-cell signaling
positive regulation of cell proliferation
Wnt signaling pathway
neurogenesis
neuron differentiation
lung development
cell proliferation in midbrain
cell fate commitment
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fibroblast proliferation
positive regulation of neurogenesis
positive regulation of sequence-specific DNA binding transcription factor activity
atrial cardiac muscle tissue morphogenesis
positive regulation of cardiac muscle cell proliferation
canonical Wnt signaling pathway
cardiac epithelial to mesenchymal transition
lung induction
positive regulation of epithelial cell proliferation involved in lung morphogenesis
labyrinthine layer blood vessel development
iris morphogenesis
mammary gland epithelium development
cellular response to retinoic acid
cellular response to transforming growth factor beta stimulus
positive regulation of canonical Wnt signaling pathway
midbrain dopaminergic neuron differentiation
canonical Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
frizzled binding
cytokine activity
protein binding
receptor agonist activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07027836.hg.1 | ENST00000265441.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027836.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027836.hg.1 | TC07001794.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000491214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000491214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000491214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | ENST00000491214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000449446.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000461427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000491214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000491214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000491214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | ENST00000491214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012368.hg.1 | NR_024047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189091.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189091.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189091.hg.1 | ENST00000265441.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189091.hg.1 | NM_003391.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189091.hg.1 | TC0700012368.hg.1 | Clariom D TC |

WNT2
Genomic Map