Assay Details
| Gene Symbol: OCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000275.2 | NP_000266.2 | 9 - 10 | 1156 | 76 | |
| XM_011521640.2 | XP_011519942.1 | 9 - 10 | 1186 | 76 | ||
| XM_017022255.1 | XP_016877744.1 | 9 - 10 | 1137 | 76 | ||
| XM_017022256.1 | XP_016877745.1 | 9 - 10 | 1634 | 76 | ||
| XM_017022258.1 | XP_016877747.1 | 9 - 10 | 1159 | 76 | ||
| XM_017022261.1 | XP_016877750.1 | 8 - 9 | 1653 | 76 | ||
| XM_017022262.1 | XP_016877751.1 | 9 - 10 | 1159 | 76 | ||
| XM_017022263.1 | XP_016877752.1 | 9 - 10 | 1159 | 76 | ||
| XM_017022264.1 | XP_016877753.1 | 9 - 10 | 1159 | 76 | ||
| XM_017022265.1 | XP_016877754.1 | 9 - 10 | 1159 | 76 | ||
| GenBank mRNA | M97901.1 | - | 2 - 3 | 185 | 76 | |
| M99564.1 | - | 9 - 10 | 1098 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001091.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009882.hg.1 | ENST00000354638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009882.hg.1 | ENST00000382996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009882.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009882.hg.1 | TC15001091.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009881.hg.1 | ENST00000353809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009881.hg.1 | ENST00000354638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009881.hg.1 | ENST00000382996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009881.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009881.hg.1 | TC15001091.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000431101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | ENST00000445578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008794.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163497.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163497.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163497.hg.1 | TC1500008794.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500163497.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163497.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163495.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163495.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163495.hg.1 | NM_000275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500163495.hg.1 | TC1500008794.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500163495.hg.1 | ENST00000353809.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163495.hg.1 | ENST00000354638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500163495.hg.1 | NM_000275.2 | Refseq |

OCA2
Genomic Map